3-(4-Nitrophenyl)isoxazole ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

N1059-200MG N1059-1G
TCIAN1059-200MGEA 75.9 EUR
TCIAN1059-200MG TCIAN1059-1G
3-(4-Nitrophenyl)isoxazole ≥96.0% (by GC)
3-(4-Nitrophenyl)isoxazole
Formule: C₉H₆N₂O₃
MDL Nummer: MFCD08446390
CAS nummer: 4264-05-5

Order Now

Specification Test Results

Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR