Neobavaisoflavone ≥97.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

N1122-25MG N1122-5MG
TCIAN1122-25MGEA 488 EUR
TCIAN1122-25MG TCIAN1122-5MG
Neobavaisoflavone ≥97.0% (by HPLC)
Neobavaisoflavone
Formule: C₂₀H₁₈O₄
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00800660
CAS nummer: 41060-15-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Carbon) 72.00 to 76.50 %
Melting point 195.0 to 199.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR