1,2,3,4,5,6,7,8-Octahydrophenanthrene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
O0021-1G
O0021-5G
TCIAO0021-1GEA
126
EUR
TCIAO0021-1G
TCIAO0021-5G
1,2,3,4,5,6,7,8-Octahydrophenanthrene ≥98.0% (by GC)
1,2,3,4,5,6,7,8-Octahydrophenanthrene
Formule:
C₁₄H₁₈ Kookpunt: 295°C |
MDL Nummer:
MFCD00058943 CAS nummer: 5325-97-3 EINECS: 226-199-6 |
Specification Test Results
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...