Solvent black 5

Leverancier: TCI EUROPE
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TCIAO0097-25GEA 20.9 EUR
TCIAO0097-25G
Solvent black 5
Solvent black 5
Formule: C₈H₁₉ClN₂O
MDL Nummer: MFCD00071316
CAS nummer: 11099-03-9

Order Now

Specification Test Results

Appearance Black powder to crystal
Lambda max. 565.0 to 569.0 nm (EtOH)
Absorbance (E1%1cm) min. 350 (EtOH, 565.0 to 569.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR