4-(2-Octylamino)diphenylamine ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
O0166-25G
O0166-500G
TCIAO0166-25GEA
17.6
EUR
TCIAO0166-25G
TCIAO0166-500G
4-(2-Octylamino)diphenylamine ≥98.0% (by GC, titration analysis)
4-(2-Octylamino)diphenylamine
Formule:
C₂₀H₂₈N₂ |
MDL Nummer:
MFCD00059441 CAS nummer: 15233-47-3 EINECS: 239-281-1 |
Specification Test Results
Appearance | Red to Dark blue to Black clear liquid to cloudy liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...