Triethyl orthobutyrate 90,0 GC_ASSAY_METHOD
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Ethyl Orthobutyrate, Orthobutyric Acid Triethyl Ester, 1,1,1-Triethoxybutane
O0173-25ML O0173-5ML
TCIAO0173-25MLEA 105 EUR
TCIAO0173-25ML TCIAO0173-5ML
Triethyl orthobutyrate 90,0 GC_ASSAY_METHOD
Triethyl orthobutyrate
Formule: C₁₀H₂₂O₃
MW: 190.28 g/mol
Kookpunt: 63°C
Vlampunt: 58°C
MDL Nummer: MFCD00059382
CAS nummer: 24964-76-9
UN: 3272
ADR: 3 III
Order Now
Specification Test Results
Appearance Colorless clear liquid
Purity (GC) min. 90.0 %