Octafluoroadipic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

O0260-25G O0260-5G
TCIAO0260-25GEA 248 EUR
TCIAO0260-25G TCIAO0260-5G
Octafluoroadipic acid ≥98.0% (by GC, titration analysis)
Octafluoroadipic acid
Formule: C₆H₂F₈O₄
MDL Nummer: MFCD00066405
CAS nummer: 336-08-3
EINECS: 206-407-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 131.0 to 136.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR