2,2'-Oxybis(ethylamine) ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

O0278-1G O0278-25G O0278-5G
TCIAO0278-1GEA 119 EUR
TCIAO0278-1G TCIAO0278-25G TCIAO0278-5G
2,2'-Oxybis(ethylamine) ≥98.0% (by GC, titration analysis)
2,2'-Oxybis(ethylamine)
Formule: C₄H₁₂N₂O
Kookpunt: 64°C
MDL Nummer: MFCD00466591
CAS nummer: 2752-17-2
EINECS: 220-395-5
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR