Oleanolic acid hydrate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

O0317-25G O0317-5G
TCIAO0317-25GEA 161 EUR
TCIAO0317-25G TCIAO0317-5G
Oleanolic acid hydrate ≥98.0% (by titrimetric analysis)
Oleanolic acid hydrate
Formule: C₃₀H₄₈O₃·xH₂O
Smeltpunt: 311°C
MDL Nummer: MFCD00064914
CAS nummer: 508-02-1
EINECS: 208-081-6
Merck Index: 14,06827
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3960eb2aad0fddb53e497b71692e0ca99d322c1&vid=37658cf8effb856ab24055e6a6fda492f5e0d6c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +74.0 to +79.0 deg (C=1, CHCl3)
Water max. 6.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR