n-Octanohydrazide ≥97.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
n-Octanoic Hydrazide
O0324-25G
O0324-5G
TCIAO0324-25GEA
101
EUR
TCIAO0324-25G
TCIAO0324-5G
n-Octanohydrazide ≥97.0% (by GC, titration analysis)
n-Octanohydrazide
Formule:
C₈H₁₈N₂O MW: 158.25 g/mol |
MDL Nummer:
MFCD00011588 CAS nummer: 6304-39-8 EINECS: 228-612-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 88.0 and le 92.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...