Orlistat ≥97.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: N-Formyl-L-leucine (1S)-1-[[(2S,3S)-3-Hexyl-4-oxo-2-oxetanyl]methyl]dodecyl Ester, (S)-(S)-1-((2S,3S)-3-Hexyl-4-oxooxetan-2-yl)tridecan-2-yl 2-formamido-4-methylpentanoate

O0381-1G O0381-100MG
TCIAO0381-1GEA 137 EUR
TCIAO0381-1G TCIAO0381-100MG
Orlistat ≥97.0% (by HPLC, total nitrogen)
Orlistat
Formule: C₂₉H₅₃NO₅
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD05662360
CAS nummer: 96829-58-2
Merck Index: 14,06869

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 43.0 to 47.0 deg-C
Specific rotation [a]20/D -33.0 to -36.0 deg (C=1, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR