3-n-Octyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 4,4,5,5-Tetramethyl-2-(3-n-octyl-2-thienyl)-1,3,2-dioxaborolan, 3-n-Octyl-2-thiopheneboronic Acid Pinacol Ester

O0410-1G O0410-5G
TCIAO0410-1GEA 80.3 EUR
TCIAO0410-1G TCIAO0410-5G
3-n-Octyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥98.0% (by GC)
3-n-Octyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene
Formule: C₁₈H₃₁BO₂S
Bewaar temperatuur: Koelkast
CAS nummer: 405165-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a620198b9df753270ba7621880c5cc2699ba50ae&vid=a620198b9df753270ba7621880c5cc2699ba50ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR