(+/-)-Octopamine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: alpha-(Aminomethyl)-4-hydroxybenzyl alcohol hydrochloride, 4-(2-Amino-1-hydroxyethyl)phenol hydrochloride, dl-Octopamine hydrochloride, 4-(2-amino-1-hydroxyethyl)phenol hydrochloride, Octopamine hydrochloride

O0413-1G O0413-5G O0413-25G
TCIAO0413-1GEA 18.7 EUR
TCIAO0413-1G TCIAO0413-5G TCIAO0413-25G
(+/-)-Octopamine hydrochloride ≥98.0% (by HPLC, total nitrogen)
(+/-)-Octopamine hydrochloride
Formule: C₈H₁₂ClNO₂
MW: 189.639 g/mol
MDL Nummer: MFCD00012881
CAS nummer: 770-05-8
EINECS: 212-216-4
Merck Index: 14,06759
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42532fc6b5475eabfc87af0ea51ed626b4437104&vid=42532fc6b5475eabfc87af0ea51ed626b4437104&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR