(S)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
4-[4-(Hexyloxy)benzoyloxy]benzoic acid (S)-2-Octyl ester
O0461-1G
O0461-5G
TCIAO0461-1GEA
71.5
EUR
TCIAO0461-1G
TCIAO0461-5G
(S)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate ≥98.0% (by GC)
(S)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate
Formule:
C₂₈H₃₈O₅ MW: 454.61 g/mol |
MDL Nummer:
MFCD01941070 CAS nummer: 87321-20-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 47.0 to 51.0 deg-C |
Specific rotation [a]20/D | +25.0 to +29.0 deg (C=5, THF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...