Sodium bis(nonafluorobutanesulphonyl)imide ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

O0492-1G O0492-5G
TCIAO0492-1GEA 62.7 EUR
TCIAO0492-1G TCIAO0492-5G
Sodium bis(nonafluorobutanesulphonyl)imide ≥97.0% (by titrimetric analysis)
Sodium bis(nonafluorobutanesulphonyl)imide
Formule: C₈F₁₈NNaO₄S₂
CAS nummer: 129135-86-0
UN: 1759
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3d2ce02cf376c176771243e5c141d69857b93f7&vid=e3d2ce02cf376c176771243e5c141d69857b93f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (Ion exchange titration) min. 97.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR