Sophocarpine monohydrate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (41S,7aS,13aR,13bR)-2,3,41,6,7,7a,8,13,13a,13b-Decahydro-1H,5H,10H-dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one hydrate, 13,14-Didehydromatridin-15-one

O0505-25MG O0505-100MG
TCIAO0505-25MGEA 93.5 EUR
TCIAO0505-25MG TCIAO0505-100MG
Sophocarpine monohydrate ≥98.0% (by HPLC)
Sophocarpine monohydrate
Formule: C₁₅H₂₂N₂O·H₂O
MDL Nummer: MFCD27923119
CAS nummer: 145572-44-7

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Elemental analysis (Nitrogen) 10.00 to 11.00 %
Specific rotation [a]20/D -23.0 to -27.0 deg (C=1, MeOH)
Water 6.0 to 9.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR