Methyl cinnamate predominantly trans 99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
|
Synoniemen:
Methyl 3-phenylpropenoate, Methyl 3-phenyl-2-propenoate
A15975.0E
A15975.22
A15975.36
A15975.0EEA
355
EUR
A15975.0E
A15975.22
A15975.36
Methyl cinnamate predominantly trans 99%
Methyl cinnamate
Purity: 99%
CAS No.: 103-26-4
Molecular Formula: C10H10O2
Formula Weight: 162.19
Flash Point: 123°(253°F)
Physical Form: Cryst solid
Density: 1.042
Refractive Index: 1.5770
Appearance: White
Melting Point: 34-37°
Boiling Point: 262-264°
Beilstein Reference: 386468
Merck Reference: 14,2299
MDL No.: MFCD00008458
Formule:
C₁₀H₁₀O₂ Kookpunt: 262…264 °C Smeltpunt: 34…37 °C Dichtheid: 1,042 Vlampunt: 123 °C (253 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008458 CAS nummer: 103-26-4 EINECS: 203-093-8 Merck Index: 14,2299 |