Pentaethylenehexamine mixture

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

P0622-25G P0622-500G
TCIAP0622-25GEA 18.7 EUR
TCIAP0622-25G TCIAP0622-500G
Pentaethylenehexamine mixture
Pentaethylenehexamine
Formule: C₁₀H₂₈N₆
Vlampunt: 185°C
MDL Nummer: MFCD00008167
CAS nummer: 4067-16-7
EINECS: 223-775-9
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) many component mixture
Amine Value 1,180 to 1,280


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR