Tris(2-butoxyethyl)phosphate ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
P0683-25G P0683-500G
TCIAP0683-25GEA 18.7 EUR
TCIAP0683-25G TCIAP0683-500G
Tris(2-butoxyethyl)phosphate ≥95.0% (by GC)
Tris(2-butoxyethyl)phosphate
Formule: C₁₈H₃₉O₇P
Kookpunt: 220°C
Vlampunt: 210°C
MDL Nummer: MFCD00009456
CAS nummer: 78-51-3
EINECS: 201-122-9
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %