Polyethylene glycol monostearate (palmitate and stearate mixture) n=∼2
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Polyoxyethylene stearate, Polyoxyl stearate
P0717-25G P0717-500G
TCIAP0717-25GEA 17.6 EUR
TCIAP0717-25G TCIAP0717-500G
Polyethylene glycol monostearate (palmitate and stearate mixture) n=∼2
Polyethylene glycol monostearate
Formule: HO(CH₂CH₂O)nOCC₁₇H₃₅
MDL Nummer: MFCD00148007
CAS nummer: 9004-99-3
Order Now
Specification Test Results
Appearance White to Almost white wax
The average value of n 1.8 to 3.2
Acid Value max. 5.0