Polyethylene glycol monostearate (palmitate and stearate mixture) n=∼4
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Polyoxyethylene stearate, Polyoxyl stearate
P0718-25G P0718-500G
TCIAP0718-25GEA 17.6 EUR
TCIAP0718-25G TCIAP0718-500G
Polyethylene glycol monostearate (palmitate and stearate mixture) n=∼4
Polyethylene glycol monostearate
Formule: HO(CH₂CH₂O)nOCC₁₇H₃₅
MDL Nummer: MFCD00148007
CAS nummer: 9004-99-3
Order Now
Specification Test Results
Appearance White to Almost white powder to lump
The average value of n 3.5 to 4.5
Acid Value max. 5.0