Polyethylene glycol monostearate (palmitate and stearate mixture) n=∼10
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P0719-25G P0719-500G
TCIAP0719-25GEA 17.6 EUR
TCIAP0719-25G TCIAP0719-500G
Polyethylene glycol monostearate (palmitate and stearate mixture) n=∼10
Polyethylene glycol monostearate
Formule: HO(CH₂CH₂O)nOCC₁₇H₃₅
MDL Nummer: MFCD00148007
CAS nummer: 9004-99-3
Order Now
Specification Test Results
Appearance White to Almost white powder to lump
The average value of n 9.0 to 13.0
Acid Value max. 1.5