Vinyl palmitate ≥96.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ)

P0848-25ML P0848-500ML
TCIAP0848-25MLEA 45.1 EUR
TCIAP0848-25ML TCIAP0848-500ML
Vinyl palmitate ≥96.0% (by GC) stabilised
Vinyl palmitate
Formule: C₁₈H₃₄O₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00059288
CAS nummer: 693-38-9
EINECS: 211-751-0

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR