Disodium phthalocyanine

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

P0973-1G P0973-5G
TCIAP0973-1GEA 30.8 EUR
TCIAP0973-1G TCIAP0973-5G
Disodium phthalocyanine
Disodium phthalocyanine
Formule: C₃₂H₁₆N₈Na₂
MDL Nummer: MFCD00066368
CAS nummer: 25476-27-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f5e1f3f8ac66aa06728da89b47f63e72fdf0b85&vid=8f5e1f3f8ac66aa06728da89b47f63e72fdf0b85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Dark red to Dark purple to Dark blue powder to crystal
Ignition residue (Sulfate) 23.0 to 25.8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR