10,12-Pentacosadiyn-1-ol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P1053-1G
P1053-100MG
TCIAP1053-1GEA
702
EUR
TCIAP1053-1G
TCIAP1053-100MG
10,12-Pentacosadiyn-1-ol ≥98.0% (by GC)
10,12-Pentacosadiyn-1-ol
Formule:
C₂₅H₄₄O |
MDL Nummer:
MFCD00059096 CAS nummer: 92266-90-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 56.0 to 61.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...