2-Phenoxypropanol ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Propylene Glycol 2-Monophenyl Ether

P1111-25G P1111-5G
TCIAP1111-25GEA 552 EUR
TCIAP1111-25G TCIAP1111-5G
2-Phenoxypropanol ≥96.0% (by GC)
2-Phenoxypropanol
Formule: C₉H₁₂O₂
Kookpunt: 244°C
MDL Nummer: MFCD00142958
CAS nummer: 4169-04-4
EINECS: 224-027-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR