Pivaloylacetonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 4,4-Dimethyl-3-oxopentanenitrile, 4,4-Dimethyl-3-oxovaleronitrile, Trimethylacetylacetonitrile

P1112-25G P1112-500G
TCIAP1112-25GEA 31.9 EUR
TCIAP1112-25G TCIAP1112-500G
Pivaloylacetonitrile ≥98.0% (by GC)
Pivaloylacetonitrile
Formule: C₇H₁₁NO
MW: 125.17 g/mol
Kookpunt: 126°C
MDL Nummer: MFCD00010208
CAS nummer: 59997-51-2
EINECS: 262-017-1
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 70.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR