Tri-o-tolyl phosphite ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Tri-o-cresyl Phosphite, Phosphorous Acid Tri-o-cresyl Ester, Phosphorous Acid Tris(2-methylphenyl) Ester, Tris(2-methylphenyl) Phosphite, Phosphorous Acid Tri-o-tolyl Ester
P1416-1G P1416-5G
TCIAP1416-1GEA 62.7 EUR
TCIAP1416-1G TCIAP1416-5G
Tri-o-tolyl phosphite ≥97.0% (by GC)
Tri-o-tolyl phosphite
Formule: C₂₁H₂₁O₃P
MW: 352.37 g/mol
Kookpunt: 238°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00014911
CAS nummer: 2622-08-4
EINECS: 220-068-7
Order Now
Specification Test Results
Purity (GC) min. 97.0 %