Pentaethylene glycol monobenzyl ether ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: BnO-PEG5-OH, 2-[2-[2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol, 1-Phenyl-2,5,8,11,14-pentaoxahexadecan-16-ol

P1435-1G P1435-5G
TCIAP1435-1GEA 141 EUR
TCIAP1435-1G TCIAP1435-5G
Pentaethylene glycol monobenzyl ether ≥95.0% (by GC)
Pentaethylene glycol monobenzyl ether
Formule: C₁₇H₂₈O₆
MDL Nummer: MFCD06797154
CAS nummer: 57671-28-0

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR