4-Phenyl-1-cyclohexene ≥96.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P1445-1G
P1445-5G
TCIAP1445-1GEA
189
EUR
TCIAP1445-1G
TCIAP1445-5G
4-Phenyl-1-cyclohexene ≥96.0% (by GC)
4-Phenyl-1-cyclohexene
Formule:
C₁₂H₁₄ Kookpunt: 235°C |
MDL Nummer:
MFCD00049188 CAS nummer: 4994-16-5 |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...