1-Pyrenecarboxaldehyde ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

P1448-1G P1448-5G
TCIAP1448-1GEA 20.9 EUR
TCIAP1448-1G TCIAP1448-5G
1-Pyrenecarboxaldehyde ≥98.0% (by GC)
1-Pyrenecarboxaldehyde
Formule: C₁₇H₁₀O
MDL Nummer: MFCD00004139
CAS nummer: 3029-19-4
EINECS: 221-196-6

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 130.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR