Palladium hydroxide on carbon (Pearlman's catalyst) (contains Pd, PdO) wetted with ca. 50% water
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
P1528-10G P1528-50G
TCIAP1528-10GEA 567 EUR
TCIAP1528-10G TCIAP1528-50G
Palladium hydroxide on carbon (Pearlman's catalyst) (contains Pd, PdO) wetted with ca. 50% water
Palladium hydroxide
Formule: H₂O₂Pd
MW: 140.435 g/mol
MDL Nummer: MFCD00064599
CAS nummer: 12135-22-7
EINECS: 235-219-2
Merck Index: 14,07060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c074b6b14514060087e2b1fb3fedb128c6990b5&vid=f3cce86307badcf330c570a4aa6762d0804b1cd7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Black powder to crystal
Drying loss 45.0 to 60.0 %