Fenol 99,5 GC_ASSAY_METHOD
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Phenylalcohol, Carbolic acid, Hydroxybenzene
P1610-25G P1610-500G
TCIAP1610-25GEA 17.6 EUR
TCIAP1610-25G TCIAP1610-500G
Fenol 99,5 GC_ASSAY_METHOD
Fenol
Formule: C₆H₅OH
MW: 94.11 g/mol
Kookpunt: 181°C
Smeltpunt: 40.8
Vlampunt: 81
MDL Nummer: MFCD00002143
CAS nummer: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1 II
Merck Index: 14,07241
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64b9cdadca882b63d679da786774304b0d5dd60f&vid=8b11625f890dd34a30243238c9e1f87f3a2a95f8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Light yellow to Light red powder to lump
Purity (GC) min. 99.5 %
Melting point 41.0 to 45.0 deg-C