4,4-Pentamethylene-2-pyrrolidone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

P1695-25G P1695-5G
TCIAP1695-25GEA 175 EUR
TCIAP1695-25G TCIAP1695-5G
4,4-Pentamethylene-2-pyrrolidone ≥98.0% (by GC)
4,4-Pentamethylene-2-pyrrolidone
Formule: C₉H₁₅NO
Kookpunt: 181°C
MDL Nummer: MFCD00177938
CAS nummer: 64744-50-9
EINECS: 451-630-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 89.0 to 93.0 deg-C
Solubility in Methanol almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR