Palladium 10% on carbon wetted with ca. 55% water useful catalyst for coupling reaction, etc.
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P1785-25G P1785-5G
TCIAP1785-25GEA 680 EUR
TCIAP1785-25G TCIAP1785-5G
Palladium 10% on carbon wetted with ca. 55% water useful catalyst for coupling reaction, etc.
Palladium
Formule: Pd
MW: 106.420 g/mol
MDL Nummer: MFCD00011167
CAS nummer: 7440-05-3
EINECS: 231-115-6
Merck Index: 14,06989
Order Now
Specification Test Results
Appearance Black powder to crystal
Palladium 9.9 to 10.1% (calcd.on dried substance)