Phenoxydiphenylphosphine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

P1843-25G P1843-5G
TCIAP1843-25GEA 549 EUR
TCIAP1843-25G TCIAP1843-5G
Phenoxydiphenylphosphine ≥98.0% (by GC)
Phenoxydiphenylphosphine
Formule: C₁₈H₁₅OP
Kookpunt: 170°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00046055
CAS nummer: 13360-92-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR