9-Phenylcarbazole-3-boronic acid (contains varying amounts of Anhydride)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
P2001-5G
P2001-25G
TCIAP2001-5GEA
115
EUR
TCIAP2001-5G
TCIAP2001-25G
9-Phenylcarbazole-3-boronic acid (contains varying amounts of Anhydride)
9-Phenylcarbazole-3-boronic acid
Formule:
C₁₈H₁₄BNO₂ |
MDL Nummer:
MFCD12196936 CAS nummer: 854952-58-2 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 107.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...