N-(2-Oxoethyl)phthalimide ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: N-(Formylmethyl)phthalimide, Phthalimidoacetaldehyde

P2010-1G P2010-5G
TCIAP2010-1GEA 73.7 EUR
TCIAP2010-1G TCIAP2010-5G
N-(2-Oxoethyl)phthalimide ≥98.0% (by total nitrogen basis)
N-(2-Oxoethyl)phthalimide
Formule: C₁₀H₇NO₃
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00023080
CAS nummer: 2913-97-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f22ff30762a2ecd3c7d6c6c901459ff1d64782ed&vid=f22ff30762a2ecd3c7d6c6c901459ff1d64782ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 112.0 to 116.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR