3-Phenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 3(5)-Phenyl-1H-pyrazole, 3-Phenylpyrazole

P2063-1G P2063-5G
TCIAP2063-1GEA 59.4 EUR
TCIAP2063-1G TCIAP2063-5G
3-Phenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
3-Phenyl-1H-pyrazole
Formule: C₉H₈N₂
Kookpunt: 314°C
MDL Nummer: MFCD00159654
CAS nummer: 2458-26-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e327d388bb4fe33ae4158d30052089675e8899bb&vid=e327d388bb4fe33ae4158d30052089675e8899bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR