Pranlukast ≥98.0% (by HPLC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
P2115-25MG P2115-100MG
TCIAP2115-25MGEA 34.1 EUR
TCIAP2115-25MG TCIAP2115-100MG
Pranlukast ≥98.0% (by HPLC)
Pranlukast
Formule: C₂₇H₂₃N₅O₄
Smeltpunt: 233°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00864631
CAS nummer: 103177-37-3
Merck Index: 14,07710
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0680804e9c13baac60b45478f2894f148c1a3608&vid=0680804e9c13baac60b45478f2894f148c1a3608&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 14.00 to 15.00 %