Phenyl methacrylate ≥97.0% (by GC) stabilised
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Stabilisatie:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
P2134-5G
P2134-25G
TCIAP2134-5GEA
49.5
EUR
TCIAP2134-5G
TCIAP2134-25G
Phenyl methacrylate ≥97.0% (by GC) stabilised
Phenyl methacrylate
Formule:
C₁₀H₁₀O₂ Kookpunt: 198°C Vlampunt: 60°C |
MDL Nummer:
MFCD00048117 CAS nummer: 2177-70-0 EINECS: 218-542-3 UN: 3272 ADR: 3,III |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...