N-Propargylmaleimide ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

P2139-25MG P2139-100MG
TCIAP2139-25MGEA 79.2 EUR
TCIAP2139-25MG TCIAP2139-100MG
N-Propargylmaleimide ≥98.0% (by HPLC, total nitrogen)
N-Propargylmaleimide
Formule: C₇H₅NO₂
Kookpunt: 88°C
MDL Nummer: MFCD12090919
CAS nummer: 209395-32-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93a6049f8496b2efc22b782b815462552d672338&vid=93a6049f8496b2efc22b782b815462552d672338&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 55.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR