Proparacaine hydrochloride ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
P2156-1G
P2156-5G
TCIAP2156-1GEA
69.3
EUR
TCIAP2156-1G
TCIAP2156-5G
Proparacaine hydrochloride ≥98.0% (by HPLC, titration analysis)
Proparacaine hydrochloride
Formule:
C₁₆H₂₆N₂O₃ |
MDL Nummer:
MFCD00083467 CAS nummer: 5875-06-9 EINECS: 227-541-7 Merck Index: 14,07807 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 182.0 to 186.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...