2-Propynylurea ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

P2171-200MG P2171-1G
TCIAP2171-200MGEA 50.6 EUR
TCIAP2171-200MG TCIAP2171-1G
2-Propynylurea ≥98.0% (by total nitrogen basis)
2-Propynylurea
Formule: C₄H₆N₂O
MDL Nummer: MFCD00052817
CAS nummer: 5221-62-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6f4ead9cdf551fd57012435f0e8d2755feafdc1&vid=a6f4ead9cdf551fd57012435f0e8d2755feafdc1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 125.0 to 130.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR