N-(2-Pyridyl)thiourea ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

P2272-5G P2272-25G
TCIAP2272-5GEA 91.3 EUR
TCIAP2272-5G TCIAP2272-25G
N-(2-Pyridyl)thiourea ≥98.0% (by HPLC, titration analysis)
N-(2-Pyridyl)thiourea
Formule: C₆H₇N₃S
MDL Nummer: MFCD00041227
CAS nummer: 14294-11-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=232075d64eb2ac6fd2a21733e958d2a9d6d0f6aa&vid=232075d64eb2ac6fd2a21733e958d2a9d6d0f6aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR