Pemetrexed ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
P2288-250MG
P2288-1G
TCIAP2288-250MGEA
59.4
EUR
TCIAP2288-250MG
TCIAP2288-1G
Pemetrexed ≥98.0% (by HPLC, titration analysis)
Pemetrexed
Formule:
C₂₀H₂₁N₅O₆ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD19105065 CAS nummer: 137281-23-3 Merck Index: 14,07077 |
Specification Test Results
Appearance | White to Blue powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Water | 2.0 to 11.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...