Polyethylene Glycol Glycidyl Lauryl ether
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P2294-25G
P2294-500G
TCIAP2294-25GEA
31.9
EUR
TCIAP2294-25G
TCIAP2294-500G
Polyethylene Glycol Glycidyl Lauryl ether
Polyethylene Glycol Glycidyl Lauryl ether
CAS nummer:
86630-59-3 |
|
Specification Test Results
Appearance | White to Almost white cloudy liquid |
Epoxy equivalent | 900 to 1,100 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...