1H,1H-Pentafluoropropyl methacrylate ≥98.0% (by GC) stabilised
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1H,1H-Pentafluoropropyl methacrylate, 2,2,3,3,3-Pentafluoropropyl Methacrylate, Methacrylic acid 2,2,2,3,3-pentafluoropropyl ester, Methacrylic acid 2,2,3,3,3-Pentafluoropropyl ester, 1H,1H-Perfluoropropyl methacrylate
Stabilisatie: Stabilised with 4-tert-butylpyrocatechol (TBC)
P2353-5G P2353-25G
TCIAP2353-5GEA 59.4 EUR
TCIAP2353-5G TCIAP2353-25G
1H,1H-Pentafluoropropyl methacrylate ≥98.0% (by GC) stabilised
1H,1H-Pentafluoropropyl methacrylate
Formule: C₇H₇F₅O₂
MW: 218.12 g/mol
Kookpunt: 46°C
Vlampunt: 29°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00039256
CAS nummer: 45115-53-5
EINECS: 256-191-8
UN: 3272
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c37a96469e726f20f53016e489af2d04f90a279d&vid=c37a96469e726f20f53016e489af2d04f90a279d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %