Piperitone (mixture of enantiomers) ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 6-Isopropyl-3-methyl-2-cyclohexen-1-one, p-Menth-1-en-3-one

P2355-25G P2355-100G
TCIAP2355-25GEA 34.1 EUR
TCIAP2355-25G TCIAP2355-100G
Piperitone (mixture of enantiomers) ≥95.0% (by GC)
Piperitone (mixture of enantiomers)
Formule: C₁₀H₁₆O
MW: 152.24 g/mol
Kookpunt: 237°C
CAS nummer: 89-81-6
EINECS: 201-942-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4d32a98497515c49d4adfdef9add4750298758a&vid=3f3b09400e20ba8b01e76906ad2a99b21735097a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR