Pamabrom ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
8-Bromotheophylline 2-Amino-2-methyl-1-propanol
P2359-1G
P2359-5G
TCIAP2359-1GEA
27.5
EUR
TCIAP2359-1G
TCIAP2359-5G
Pamabrom ≥98.0% (by HPLC, titration analysis)
Pamabrom
Formule:
C₁₁H₁₈BrN₅O₃ Smeltpunt: 295°C |
MDL Nummer:
MFCD00867328 CAS nummer: 606-04-2 EINECS: 210-103-4 Merck Index: 14,07000 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...